2-Amino-3-chlor-5-(trifluormethyl)pyridin

Lieferant: Thermo Scientific

SB01178.1
MAYBSB01178.1EA 0 EUR
MAYBSB01178.1
2-Amino-3-chlor-5-(trifluormethyl)pyridin
2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Formel: C₆H₄ClF₃N₂
Molecular Weight: 196,5600 g/mol
MDL: MFCD00042154
CAS-Nummer: 79456-26-1
EINECS: 401-670-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR