3,5-Dimethylthiophenol
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethylbenzol-1-thiol, 1,3-Dimethyl-5-mercaptobenzol, 3,5-Dimethylphenylmercaptan, 3,5-Dimethylbenzolthiol, 5-Mercapto-m-xylol
SB01213DA
SB01213EA
SB01213.50
MAYBSB01213DAEA
0
EUR
MAYBSB01213DA
MAYBSB01213EA
MAYBSB01213.50
3,5-Dimethylthiophenol
3,5-Dimethylthiophenol
Formel:
C₈H₁₀S Molecular Weight: 138.23 g/mol Siedepunkt: 96 Dichte: 1.02 g/cm³ (20 °C) |
MDL:
MFCD00040226 CAS-Nummer: 38360-81-5 EINECS: 253-901-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...