2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol
Lieferant: Thermo Scientific
SB01223EA
SB01223EE
MAYBSB01223EAEA
295
EUR
MAYBSB01223EA
MAYBSB01223EE
2,4-Dimethyl-6-(2-methyl-2-propanyl)phenol
6-tert-Butyl-2,4-xylenol
Formel:
C₁₂H₁₈O |
MDL:
MFCD00002234 CAS-Nummer: 1879-09-0 EINECS: 217-533-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...