2,3-Dihydrobenzo[b]furan

Lieferant: Thermo Scientific

SB01482.50
MAYBSB01482.50EA 0 EUR
MAYBSB01482.50
2,3-Dihydrobenzo[b]furan
2,3-Dihydrobenzo[b]furan
Formel: C₈H₈O
Molecular Weight: 120,15 g/mol
MDL: MFCD00005855
CAS-Nummer: 496-16-2
EINECS: 207-817-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR