Ammoniumthiocyanat
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
SB01587.250
SB01587.1
CHMPSB01587.250EA
0
EUR
CHMPSB01587.250
MAYBSB01587.1
Ammoniumthiocyanat
Ammoniumthiocyanat
Formel:
NH₄SCN Schmelzpunkt: 150 °C Dichte: 1,305 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...