3-(Trifluormethyl)phenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
α,α,α-Trifluoro-m-tolyl isothiocyanate
SB01602.1
SB01602.10
SB01602.50
MAYBSB01602.1EA
0
EUR
MAYBSB01602.1
MAYBSB01602.10
MAYBSB01602.50
3-(Trifluormethyl)phenylisothiocyanat
3-(Trifluormethyl)phenylisothiocyanat
Formel:
C₈H₄F₃NS Siedepunkt: 206…208 °C (1013 hPa) Dichte: 1,33 g/cm³ (20 °C) |
MDL:
MFCD00004806 CAS-Nummer: 1840-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...