4-Butylanilin
Lieferant: Thermo Scientific
Synonyme:
4-Butylbenzenamine
SB01608-1
MAYBSB01608-1EA
0
EUR
MAYBSB01608-1
4-Butylanilin
4-Butylanilin
Formel:
C₁₀H₁₅N Molecular Weight: 149,24 g/mol Siedepunkt: 242 °C (1013 hPa) Schmelzpunkt: –14 °C Dichte: 0,945 g/cm³ (20 °C) |
MDL:
MFCD00007925 CAS-Nummer: 104-13-2 EINECS: 203-177-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...