α,4-Dibrom-2-fluortoluol
Lieferant: Thermo Scientific
Synonyme:
4-Brom-2-fluorbenzylbromid, 1-Brom-4-(brommethyl)-3-fluorbenzol
SB01611-1
MAYBSB01611-1EA
0
EUR
MAYBSB01611-1
α,4-Dibrom-2-fluortoluol
α,4-Dibrom-2-fluortoluol
Formel:
C₇H₅Br₂F Molecular Weight: 267,92 g/mol Siedepunkt: 126 °C (19 mmHg) Schmelzpunkt: 30 °C |
MDL:
MFCD00055467 CAS-Nummer: 76283-09-5 EINECS: 278-412-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...