3,5-Bis(trifluormethyl)phenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 1,3-Bis(trifluormethyl)-5-isothiocyanatobenzol

SB01673.1 SB01673EA SB01673.50
MAYBSB01673.1EA 0 EUR
MAYBSB01673.1 MAYBSB01673EA MAYBSB01673.50
3,5-Bis(trifluormethyl)phenylisothiocyanat
3,5-Bis(trifluormethyl)phenylisothiocyanat
Formel: C₉H₃F₆NS
Siedepunkt: 94…95 °C (19 mmHg)
Dichte: 1,48 g/cm³ (20 °C)
MDL: MFCD00040838
CAS-Nummer: 23165-29-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR