3,5-Bis(trifluormethyl)phenylisothiocyanat
Lieferant: Thermo Scientific
Synonyme:
1,3-Bis(trifluormethyl)-5-isothiocyanatobenzol
SB01673.1
SB01673EA
SB01673.50
MAYBSB01673.1EA
0
EUR
MAYBSB01673.1
MAYBSB01673EA
MAYBSB01673.50
3,5-Bis(trifluormethyl)phenylisothiocyanat
3,5-Bis(trifluormethyl)phenylisothiocyanat
Formel:
C₉H₃F₆NS Siedepunkt: 94…95 °C (19 mmHg) Dichte: 1,48 g/cm³ (20 °C) |
MDL:
MFCD00040838 CAS-Nummer: 23165-29-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...