(3,5-Difluorphenyl)acetonitril
Lieferant: Thermo Scientific
SB01685.1
SB01685EA
MAYBSB01685.1EA
0
EUR
MAYBSB01685.1
MAYBSB01685EA
(3,5-Difluorphenyl)acetonitril
(3,5-Difluorphenyl)acetonitril
Formel:
C₈H₅F₂N Siedepunkt: 221 °C (1013 hPa) Schmelzpunkt: 34…36 °C |
MDL:
MFCD00061278 CAS-Nummer: 122376-76-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...