(2-Hydroxyphenyl)essigsäure
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxyphenylacetic acid
SB01717.1
MAYBSB01717.1EA
0
EUR
MAYBSB01717.1
(2-Hydroxyphenyl)essigsäure
(2-Hydroxyphenyl)essigsäure
Formel:
(C₆H₄OH)CH₂COOH Molecular Weight: 152,15 g/mol Siedepunkt: 328 °C (1013 hPa) Schmelzpunkt: 145…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004323 CAS-Nummer: 614-75-5 EINECS: 210-393-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...