3-Chlor-2-methylphenylisocyanat
Lieferant: Thermo Scientific
Synonyme:
Isocyansäure(3-chlor-2-methylphenyl)ester
SB01803.1
SB01803.10
SB01803.50
MAYBSB01803.1EA
0
EUR
MAYBSB01803.1
MAYBSB01803.10
MAYBSB01803.50
3-Chlor-2-methylphenylisocyanat
3-Chlor-2-methylphenylisocyanat
Formel:
C₈H₆ClNO Siedepunkt: 78…80 °C (1013 hPa) Dichte: 1,247 g/cm³ (20 °C) |
MDL:
MFCD00037036 CAS-Nummer: 40397-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...