Ethyl-3-ethoxyacrylat

Lieferant: Thermo Scientific

Synonyme: 3-Ethoxyacrylic acid ethyl ester, Ethyl 3-Ethoxyacrylate, Ethyl 3-Ethoxy-2-Propenoate, Ethyl 3-ethoxyacrylate

SB01829DA SB01829.50
MAYBSB01829DAEA 0 EUR
MAYBSB01829DA MAYBSB01829.50
Ethyl-3-ethoxyacrylat
Ethyl-3-ethoxyacrylat
Formel: C₇H₁₂O₃
Molecular Weight: 144,17 g/mol
MDL: MFCD00009863
CAS-Nummer: 1001-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR