2-Fluorphenylisocyanat
Lieferant: Thermo Scientific
Synonyme:
1-Fluoro-2-isocyanatobenzene
SB01843DA
SB01843.10
MAYBSB01843DAEA
0
EUR
MAYBSB01843DA
MAYBSB01843.10
2-Fluorphenylisocyanat
2-Fluorphenylisocyanat
Formel:
C₇H₄FNO Molecular Weight: 137,11 g/mol Siedepunkt: 41…44 °C (1 mmHg) Dichte: 1,222 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00001996 CAS-Nummer: 16744-98-2 EINECS: 240-804-0 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...