4-Chlor-2,5-dimethoxyphenylisothiocyanat

Lieferant: Thermo Scientific

SB01846.50
MAYBSB01846.50EA 0 EUR
MAYBSB01846.50
4-Chlor-2,5-dimethoxyphenylisothiocyanat
4-Chlor-2,5-dimethoxyphenylisothiocyanat
Formel: C₉H₈ClNO₂S
MDL: MFCD00046809
CAS-Nummer: 306935-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR