1,3,5-Trifluorbenzol
Lieferant: Thermo Scientific
SB01896.1
SB01896.50
MAYBSB01896.1EA
0
EUR
MAYBSB01896.1
MAYBSB01896.50
1,3,5-Trifluorbenzol
1,3,5-Trifluorobenzene
Formel:
C₆H₃F₃ Molecular Weight: 132.09 g/mol Siedepunkt: 75 Dichte: 1.277 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000333 CAS-Nummer: 372-38-3 EINECS: 206-751-2 UN: 2387 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...