4'-Hydroxy-3'-methylacetophenon

Lieferant: Thermo Scientific

SB01922.1
MAYBSB01922.1EA 0 EUR
MAYBSB01922.1
4'-Hydroxy-3'-methylacetophenon
4'-Hydroxy-3'-methylacetophenon
Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
MDL: MFCD00002231
CAS-Nummer: 876-02-8
EINECS: 212-880-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR