3-(Trifluormethyl)zimtsäure
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Trifluormethyl)cinnamsäure, 3-(3-(Trifluormethyl)phenyl)propensäure, 3-(m-(Trifluormethyl)phenyl)propensäure
SB01924.1
SB01924EA
SB01924.50
MAYBSB01924.1EA
0
EUR
MAYBSB01924.1
MAYBSB01924EA
MAYBSB01924.50
3-(Trifluormethyl)zimtsäure
3-(Trifluormethyl)zimtsäure
Formel:
C₁₀H₇F₃O₂ Molecular Weight: 216.16 g/mol Siedepunkt: 292 Schmelzpunkt: 130…131 °C |
MDL:
MFCD00004393 CAS-Nummer: 779-89-5 EINECS: 212-301-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...