4-(Trifluormethylthio)phenol

Lieferant: Thermo Scientific

SB01940DA SB01940EA
MAYBSB01940DAEA 0 EUR
MAYBSB01940DA MAYBSB01940EA
4-(Trifluormethylthio)phenol
4-(Trifluormethylthio)phenol
Formel: C₇H₅F₃OS
MDL: MFCD00036035
CAS-Nummer: 461-84-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR