(R)-(+)-1,2-Epoxypropan

Lieferant: Thermo Scientific

SB02051-1
MAYBSB02051-1EA 0 EUR
MAYBSB02051-1
(R)-(+)-1,2-Epoxypropan
(R)-(+)-1,2-Epoxypropan
Formel: C₃H₆O
Dichte: 0,829 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066211
CAS-Nummer: 15448-47-2
UN: 1280
ADR: 3,I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR