4-Fluor-3-nitrobenzaldehyd

Lieferant: Thermo Scientific

SB02056DA
MAYBSB02056DAEA 0 EUR
MAYBSB02056DA
4-Fluor-3-nitrobenzaldehyd
4-Fluoro-3-nitrobenzaldehyde
Formel: C₇H₄FNO₃
Molecular Weight: 169,11 g/mol
MDL: MFCD01861388
CAS-Nummer: 42564-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR