2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat

Lieferant: Thermo Scientific

SB02090.250
MAYBSB02090.250EA 0 EUR
MAYBSB02090.250
2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat
2,4-Di-tert-butoxy-5-pyrimidinboronsäure Hydrat
Formel: C₁₂H₂₁BN₂O₄·H₂O
MDL: MFCD02180675
CAS-Nummer: 306935-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR