3,5-Bis(trifluormethyl)benzaldehyd
Lieferant: Thermo Scientific
SEW00692.1
MAYBSEW00692.1EA
0
EUR
MAYBSEW00692.1
3,5-Bis(trifluormethyl)benzaldehyd
3,5-Bis(trifluormethyl)benzaldehyd
Formel:
C₉H₄F₆O Molecular Weight: 242,12 g/mol Siedepunkt: 47…48 °C (0,2 mmHg) Schmelzpunkt: –2 °C Dichte: 1,46 g/cm³ (20 °C) |
MDL:
MFCD00010206 CAS-Nummer: 401-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...