2-Chlor-4-fluorbenzaldehyd
Lieferant: Thermo Scientific
|
Achtung
|
SEW01013.10
MAYBSEW01013.10EA
0
EUR
MAYBSEW01013.10
2-Chlor-4-fluorbenzaldehyd
2-Chloro-4-fluorobenzaldehyde
Formel:
C₇H₄ClFO Molecular Weight: 158,56 g/mol Siedepunkt: 205 °C (1013 hPa) Schmelzpunkt: 60 °C |
MDL:
MFCD00042527 CAS-Nummer: 84194-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...