3-Chlor-4-iodbenzotrifluorid

Lieferant: Thermo Scientific

SEW01265.10
MAYBSEW01265.10EA 0 EUR
MAYBSEW01265.10
3-Chlor-4-iodbenzotrifluorid
3-Chlor-4-iodbenzotrifluorid
Formel: C₇H₃ClF₃I
MDL: MFCD00042562
CAS-Nummer: 141738-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR