2-Brom-6-chlor-4-(trifluormethyl)anilin

Lieferant: Thermo Scientific

Synonyme: 4-Amino-3-brom-5-chlor-α,α,α-trifluortoluol, 4-Amino-3-brom-5-chlorbenzotrifluorid

SEW01272DA SEW01272.10
MAYBSEW01272DAEA 0 EUR
MAYBSEW01272DA MAYBSEW01272.10
2-Brom-6-chlor-4-(trifluormethyl)anilin
2-Brom-6-chlor-4-(trifluormethyl)anilin
Formel: C₇H₄BrClF₃N
Molecular Weight: 274,47 g/mol
Siedepunkt: 71…72 °C (0,25 mmHg)
Schmelzpunkt: 29…32 °C
MDL: MFCD00042180
CAS-Nummer: 109919-26-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR