2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
2-Chlor-4-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Chlor-4-(trifluormethyl)benzolsulfochlorid, 2-Chlor-4-(trifluormethyl)benzolsulfonsäurechlorid
SEW01287.250
SEW01287DA
MAYBSEW01287.250EA
0
EUR
MAYBSEW01287.250
MAYBSEW01287DA
2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃Cl₂F₃O₂S Siedepunkt: 70…71 °C (0,5 mmHg) |
MDL:
MFCD00052912 CAS-Nummer: 175205-54-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...