3-Brom-2-thenoesäure
Lieferant: Thermo Scientific
Synonyme:
3-Bromthiophen-2-carbonsäure, 3-Brom-2-thiophencarbonsäure
SEW01838DA
MAYBSEW01838DAEA
0
EUR
MAYBSEW01838DA
3-Brom-2-thenoesäure
3-Brom-2-thenoesäure
Formel:
C₅H₃BrO₂S Molecular Weight: 207,05 g/mol |
MDL:
MFCD00052291 CAS-Nummer: 7311-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...