3-Brom-2-thiophencarbaldehyd

Lieferant: Thermo Scientific

Synonyme: 3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene

SEW01928.250 SEW01928DA
MAYBSEW01928.250EA 0 EUR
MAYBSEW01928.250 MAYBSEW01928DA
3-Brom-2-thiophencarbaldehyd
3-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191,05 g/mol
MDL: MFCD00126680
CAS-Nummer: 930-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR