3,4-Dimethylthiophen

Lieferant: Thermo Scientific

SEW02820.250 SEW02820.1
MAYBSEW02820.250EA 0 EUR
MAYBSEW02820.250 MAYBSEW02820.1
3,4-Dimethylthiophen
3,4-Dimethylthiophene
Formel: C₆H₈S
Molecular Weight: 112.1900 g/mol
MDL: MFCD01764731
CAS-Nummer: 632-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR