4-Amino-2-methylpyrimidin-5-carbosäurenitril

Lieferant: Thermo Scientific

Synonyme: 4-Amino-5-cyano-2-methylpyrimidine

SEW03366.250
MAYBSEW03366.250EA 0 EUR
MAYBSEW03366.250
4-Amino-2-methylpyrimidin-5-carbosäurenitril
4-Amino-2-methylpyrimidin-5-carbosäurenitril
Formel: C₆H₆N₄
Molecular Weight: 134,14 g/mol
MDL: MFCD00084875
CAS-Nummer: 698-29-3
EINECS: 211-814-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR