2,4-Dibrom-6-(trifluormethyl)anilin

Lieferant: Thermo Scientific

SEW04149.1 SEW04149.10 SEW04149.50
MAYBSEW04149.1EA 0 EUR
MAYBSEW04149.1 MAYBSEW04149.10 MAYBSEW04149.50
2,4-Dibrom-6-(trifluormethyl)anilin
2,4-Dibrom-6-(trifluormethyl)anilin
Formel: C₇H₄Br₂F₃N
MDL: MFCD00178769
CAS-Nummer: 71757-14-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR