Tributyl(4-fluorphenyl)stannan

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Fluoro-4-(tributylstannyl)benzene

SP00184DA SP00184.10
MAYBSP00184DAEA 0 EUR
MAYBSP00184DA MAYBSP00184.10
Tributyl(4-fluorphenyl)stannan
Tributyl(4-fluorphenyl)stannan
Formel: C₁₈H₃₁FSn
MDL: MFCD01313817
CAS-Nummer: 17151-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR