(2,4-Dichlorphenoxy)acetonitril

Lieferant: Thermo Scientific

SPB00370.50
MAYBSPB00370.50EA 0 EUR
MAYBSPB00370.50
(2,4-Dichlorphenoxy)acetonitril
(2,4-Dichlorphenoxy)acetonitril
Formel: C₈H₅Cl₂NO
Molecular Weight: 202.04 g/mol
MDL: MFCD00019826
CAS-Nummer: 3956-63-6
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR