2,4'-Dichloracetanilid

Lieferant: Thermo Scientific

Synonyme: N-(4-Chlorophenyl)-2-chloroacetamide, 2-Chloro-N-(4-chlorophenyl)acetamide, 2,4′-Dichloroacetanilide

SPB00454.50
MAYBSPB00454.50EA 0 EUR
MAYBSPB00454.50
2,4'-Dichloracetanilid
2,4'-Dichloracetanilid
Formel: C₈H₇Cl₂NO
Molecular Weight: 204,0533 g/mol
MDL: MFCD00018904
CAS-Nummer: 3289-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR