(3,5-Dichlorphenoxy)acetonitril
Lieferant: Thermo Scientific
SPB01308.1
SPB01308.10
MAYBSPB01308.1EA
0
EUR
MAYBSPB01308.1
MAYBSPB01308.10
(3,5-Dichlorphenoxy)acetonitril
(3,5-Dichlorphenoxy)acetonitril
Formel:
C₈H₅Cl₂NO Molecular Weight: 202.04 g/mol |
MDL:
MFCD00052707 CAS-Nummer: 103140-12-1 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...