4-Chlor-N-ethyl-2-nitroanilin

Lieferant: Thermo Scientific

SPB02522
MAYBSPB02522EA 0 EUR
MAYBSPB02522
4-Chlor-N-ethyl-2-nitroanilin
4-Chlor-N-ethyl-2-nitroanilin
Formel: C₈H₉ClN₂O₂
MDL: MFCD00024323
CAS-Nummer: 28491-95-4
EINECS: 249-055-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR