6-Chlornicotinonitril

Lieferant: Thermo Scientific

SPB04745.0003 SPB04745.250 SPB04745.1
MAYBSPB04745.0003EA 0 EUR
MAYBSPB04745.0003 MAYBSPB04745.250 MAYBSPB04745.1
6-Chlornicotinonitril
6-Chlornicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00084941
CAS-Nummer: 33252-28-7
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR