6-Methoxynicotinonitril

Lieferant: Thermo Scientific

Synonyme: 2-Methoxypyridine-5-carbonitrile

SPB04785DA SPB04785.10
MAYBSPB04785DAEA 0 EUR
MAYBSPB04785DA MAYBSPB04785.10
6-Methoxynicotinonitril
6-Methoxynicotinonitril
Formel: C₇H₆N₂O
Molecular Weight: 134.1400 g/mol
MDL: MFCD00084981
CAS-Nummer: 15871-85-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR