3,4-Dichlorbenzaldehydoxim
Lieferant: Thermo Scientific
Synonyme:
3,4-Dichlorobenzaldoxime
SPB04935.1
MAYBSPB04935.1EA
0
EUR
MAYBSPB04935.1
3,4-Dichlorbenzaldehydoxim
3,4-Dichlorbenzaldehydoxim
Formel:
C₇H₅Cl₂NO |
MDL:
MFCD00017592 CAS-Nummer: 5331-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...