2,6-Dibrom-4-(trifluormethoxy)anilin

Lieferant: Thermo Scientific

SPB05517.1 SPB05517.10
MAYBSPB05517.1EA 0 EUR
MAYBSPB05517.1 MAYBSPB05517.10
2,6-Dibrom-4-(trifluormethoxy)anilin
2,6-Dibrom-4-(trifluormethoxy)anilin
Formel: C₇H₄Br₂F₃NO
MDL: MFCD00153113
CAS-Nummer: 88149-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR