2-Brom-5-(trifluormethyl)pyridin
Lieferant: Thermo Scientific
SPB05624DA
MAYBSPB05624DAEA
0
EUR
MAYBSPB05624DA
2-Brom-5-(trifluormethyl)pyridin
2-Bromo-5-(trifluoromethyl)pyridine
Formel:
C₆H₃BrF₃N Molecular Weight: 226 g/mol |
MDL:
MFCD00153086 CAS-Nummer: 50488-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...