2-Chlorisonicotinonitril

Lieferant: Thermo Scientific

SPB06434.1 SPB06434.50
MAYBSPB06434.1EA 0 EUR
MAYBSPB06434.1 MAYBSPB06434.50
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00174318
CAS-Nummer: 33252-30-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR