2-Chlor-6-fluorbenzoylchlorid
Lieferant: Thermo Scientific
Synonyme:
2-Chlor-6-fluorbenzol-1-carbonylchlorid
SPB06439.10
MAYBSPB06439.10EA
0
EUR
MAYBSPB06439.10
2-Chlor-6-fluorbenzoylchlorid
2-Chlor-6-fluorbenzoylchlorid
Formel:
C₇H₃Cl₂FO Siedepunkt: 227 °C (1013 hPa) Schmelzpunkt: 57 °C |
MDL:
MFCD00178746 CAS-Nummer: 79455-63-3 EINECS: 279-162-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...