2-Chlorisonicotinsäure

Lieferant: Thermo Scientific

Synonyme: 2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure

SPB06649.0010 SPB06649.1 SPB06649EA
MAYBSPB06649.0010EA 0 EUR
MAYBSPB06649.0010 MAYBSPB06649.1 MAYBSPB06649EA
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel: C₆H₄ClNO₂
MDL: MFCD00191402
CAS-Nummer: 6313-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR