1-(3,5-Dichlor-4-pyridinyl)piperazin

Lieferant: Thermo Scientific

SPB06834DA
MAYBSPB06834DAEA 0 EUR
MAYBSPB06834DA
1-(3,5-Dichlor-4-pyridinyl)piperazin
1-(3,5-Dichlor-4-pyridinyl)piperazin
Formel: C₉H₁₁Cl₂N₃
Molecular Weight: 232,11 g/mol
MDL: MFCD00221400
CAS-Nummer: 175277-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR