4-(Chlormethyl)-3-(2,6-dichlorphenyl)-5-methylisoxazol

Lieferant: Thermo Scientific

SPB07543.1
MAYBSPB07543.1EA 0 EUR
MAYBSPB07543.1
4-(Chlormethyl)-3-(2,6-dichlorphenyl)-5-methylisoxazol
4-(Chloromethyl)-3-(2,6-dichlorophenyl)-5-methylisoxazole
Formel: C₁₁H₈Cl₃NO
MDL: MFCD00833265
CAS-Nummer: 303225-22-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR