ω-Brom-3-cyanacetophenon

Lieferant: Thermo Scientific

SPB07593.0003 SPB07593DA
MAYBSPB07593.0003EA 0 EUR
MAYBSPB07593.0003 MAYBSPB07593DA
ω-Brom-3-cyanacetophenon
ω-Brom-3-cyanacetophenon
Formel: C₉H₆BrNO
MDL: MFCD00833267
CAS-Nummer: 50916-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR