2-Chlor-6-fluorbenzaldehydoxim

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-6-fluorobenzaldoxime

SPB08294.50
MAYBSPB08294.50EA 0 EUR
MAYBSPB08294.50
2-Chlor-6-fluorbenzaldehydoxim
2-Chlor-6-fluorbenzaldehydoxim
Formel: C₇H₅ClFNO
MDL: MFCD00837704
CAS-Nummer: 443-33-4
EINECS: 207-135-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR